2,3'-Difluorobenzophenone ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

210741GM
100293-614EA 210.54 USD
100293-614
2,3'-Difluorobenzophenone ≥97%
2,3'-Difluorobenzophenone
Formula: C₁₃H₈F₂O
MDL Number: MFCD00017914
CAS Number: 58139-11-0
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/72/66/21287266.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR