2,3-Difluorobenzamide ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

0933215GM
100283-300EA 124.23 USD
100283-300
2,3-Difluorobenzamide ≥97%
2,3-Difluorobenzamide
Formula: C₇H₅F₂NO
Melting Pt: 114…119 °C
MDL Number: MFCD00061137
CAS Number: 18355-75-4
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR