2,4-Dichloro-6,7-dimethoxyquinazoline ≥98%, crystalline solid

Supplier: INDOFINE CHEMICAL COMPANY MS

250161GM
100267-880EA 58.85 USD
100267-880
2,4-Dichloro-6,7-dimethoxyquinazoline ≥98%, crystalline solid
2,4-Dichloro-6,7-dimethoxyquinazoline

  • Pale yellow crystalline solid
  • Store in a cool, dry place

Formula: C₁₀H₈Cl₂N₂O₂
MW: 259.0887 g/mol
Melting Pt: 162…164 °C
MDL Number: MFCD00051733
CAS Number: 27631-29-4
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/57/78/21285778.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR