2,5-Dibromo-3,4,6-trifluoroaniline ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

270971GM
100297-460EA 281.15 USD
100297-460
2,5-Dibromo-3,4,6-trifluoroaniline ≥97%
2,5-Dibromo-3,4,6-trifluoroaniline
Formula: C₆H₂Br₂F₃N
MDL Number: MFCD01862003
CAS Number: 232267-32-2
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/74/77/21287477.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR