Daidzin ≥96%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 7-(beta-D-Glucopyranosyloxy)-4'-hydroxyisoflavone

0210965MG
100264-674EA 128.15 USD
100264-674
Daidzin ≥96%, powder
Daidzin

  • Off white powder
  • Store in a cool, dry place

Formula: C₂₁H₂₀O₉
MW: 416.38 g/mol
Melting Pt: 230…238 °C
MDL Number: MFCD00017466
CAS Number: 552-66-9
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/56/18/21285618.jpg https://us.vwr.com/stibo/web/std.lang.all/56/19/21285619.jpg https://us.vwr.com/stibo/web/std.lang.all/56/20/21285620.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR