Cytosine ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 6-Aminopyrimidin-2(1H)-one4-Amino-2-pyrimidinol

0270321GM
100266-530EA 71.92 USD
100266-530
Cytosine ≥98%
Cytosine
Formula: C₄H₅N₃O
MW: 111.1 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00006034
CAS Number: 71-30-7
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/57/20/21285720.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR