Cytochalasin D ≥98% (by TLC), powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 7(S),18(R)-Dihydroxy-16(S),18-dimethyl-10-phenyl[11]cytochalasa-6(12),13(E),19(E)-triene-1,17-dione 21(R)-acetate

0215641MG
100269-800EA 452.46 USD
100269-800
Cytochalasin D ≥98% (by TLC), powder
Cytochalasin D
Formula: C₃₀H₃₇NO₆
Melting Pt: 255…260 °C
Storage Temperature: Freezer
MDL Number: MFCD00077706
CAS Number: 22144-77-0
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/59/30/21285930.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR