(S)-2-Amino-2-cyclohexylacetic acid hydrochloride, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

0410691GM
100278-680EA 128.15 USD
100278-680
(S)-2-Amino-2-cyclohexylacetic acid hydrochloride, powder
(S)-2-Amino-2-cyclohexylacetic acid hydrochloride

  • White powder
  • Store at 0-8 °C

Formula: C₈H₁₆ClNO₂
MW: 193.671 g/mol
MDL Number: MFCD00050539
CAS Number: 191611-20-8
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR