H-D-Chg-OH·HCI ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: H-D-Chg-OH.HCl

0410681GM
100278-678EA 104.62 USD
100278-678
H-D-Chg-OH·HCI ≥99%, powder
H-D-Chg-OH·HCI

  • White powder
  • Store at 0-8 °C

Formula: C₈H₁₅NO₂·HCl
MW: 193.7 g/mol
MDL Number: MFCD00067543
CAS Number: 61367-40-6
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR