β-Cyclodextrine hydrate
Supplier: INDOFINE CHEMICAL COMPANY MS
02443925GM
100266-288EA
188.31
USD
100266-288
β-Cyclodextrine hydrate
β-Cyclodextrine hydrate
Formula:
C₄₂H₇₀O₃₅·H₂O Melting Pt: 288 °C (dec.) |
MDL Number:
MFCD00150811 CAS Number: 68168-23-0 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...