3-Cyano-7-ethoxycoumarin ≥98%, Powder

Supplier: INDOFINE CHEMICAL COMPANY MS

19033250MG
100265-078EA 41.85 USD
100265-078
3-Cyano-7-ethoxycoumarin ≥98%, Powder
3-Cyano-7-ethoxycoumarin

  • Deep pink fluffy powder
  • Store in a cool, dry place

Formula: C₁₂H₉NO₃
MW: 215.21 g/mol
Melting Pt: 210…212 °C
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/56/35/21285635.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR