3,3',4',5,7-Pentahydroxyflavylium chloride ≥96% (by HPLC), powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Cyanidin chloride

020909S10MG
100268-618EA 245.85 USD
100268-618
3,3',4',5,7-Pentahydroxyflavylium chloride ≥96% (by HPLC), powder
3,3',4',5,7-Pentahydroxyflavylium chloride

  • Violet powder
  • To be kept in a desiccator and away from light

Formula: C₁₅H₁₁O₆Cl
MW: 322.7 g/mol
MDL Number: MFCD00017582
CAS Number: 528-58-5
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/58/58/21285858.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR