Coniferyl alcohol ≥95% (by HPLC)

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 4-(3-Hydroxyprop-1-en-1-yl)-2-methoxyphenol

026028100MG
100268-970EA 295.02 USD
100268-970
Coniferyl alcohol ≥95% (by HPLC)
Coniferyl alcohol
Formula: C₁₀H₁₂O₃
MW: 180.20
MDL Number: MFCD00002922
CAS Number: 458-35-5
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/58/81/21285881.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR