Cinnamyl alcohol ≥98%, solid

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Cinnamic alcohol3-Phenylprop-2-en-1-ol

026025100GM
100263-762EA 32.69 USD
100263-762
Cinnamyl alcohol ≥98%, solid
Cinnamyl alcohol

  • White, fused solid
  • Store in a cool, dry place

Formula: C₉H₁₀O
MW: 134.18 g/mol
Melting Pt: 32…35 °C
MDL Number: MFCD00002921
CAS Number: 104-54-1
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/55/82/21285582.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR