Cholesterol acetate ≥97%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Acetic Acid Cholesterol EsterAcetyl Cholesterol

02356225GM
100269-878EA 338.69 USD
100269-878
Cholesterol acetate ≥97%
Cholesterol acetate
Formula: C₂₉H₄₈O₂
MW: 428.70 g/mol
Melting Pt: 112…114 °C
MDL Number: MFCD00003636
CAS Number: 604-35-3
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/59/34/21285934.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR