4-Chloro-6-(trifluoromethyl)quinoline ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

265081GM
100288-832EA 232.77 USD
100288-832
4-Chloro-6-(trifluoromethyl)quinoline ≥98%
4-Chloro-6-(trifluoromethyl)quinoline
Formula: C₁₀H₅ClF₃N
MW: 231.61 g/mol
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/70/36/21287036.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR