2-Chloro-5-(trifluoromethyl)benzoic acid ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 2-Chloro-5-trifluoromethylbenzoic acid6-Chloro-α,α,α-trifluoro-m-toluic acid

0932735GM
100274-610EA 111.15 USD
100274-610
2-Chloro-5-(trifluoromethyl)benzoic acid ≥98%
2-Chloro-5-(trifluoromethyl)benzoic acid
Formula: C₈H₄ClF₃O₂
MW: 224.57 g/mol
Melting Pt: 93…96 °C
MDL Number: MFCD00061094
CAS Number: 657-06-7
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/61/96/21286196.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR