3-Chloro-4-(trifluoromethoxy)benzyl bromide ≥98%, liquid

Supplier: INDOFINE CHEMICAL COMPANY MS

072571GM
100282-380EA 170 USD
100282-380
3-Chloro-4-(trifluoromethoxy)benzyl bromide ≥98%, liquid
3-Chloro-4-(trifluoromethoxy)benzyl bromide

  • Colorless liquid
  • Store in cool, dry place

Formula: C₈H₅BrClF₃O
MDL Number: MFCD01631560
CAS Number: 261763-18-2
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/68/03/21286803.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR