6-Chloro-3-indolyl-β-D-galactopyranoside ≥98% (by TLC), powder
Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms:
Rose-Gal
, Salmon-Gal
MB1026100MG
100268-414EA
52.31
USD
100268-414
6-Chloro-3-indolyl-β-D-galactopyranoside ≥98% (by TLC), powder
6-Chloro-3-indolyl-beta-D-galactopyranoside
- White powder
- Store at 0-8 °C
Formula:
C₁₄H₁₆ClNO₆ MW: 329.733 g/mol Melting Pt: 188…190 °C |
MDL Number:
MFCD00467206 CAS Number: 138182-21-5 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...