6-Chloro-4-hydroxycoumarin ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 6-Chloro-4-hydroxy-2H-chromen-2-one

17054250MG
100265-016EA 28.77 USD
100265-016
6-Chloro-4-hydroxycoumarin ≥98%, powder
6-Chloro-4-hydroxycoumarin

  • White powder
  • Store in a cool, dry place

Formula: C₉H₅ClO₃
MW: 196.59 g/mol
Melting Pt: 287…288 °C
MDL Number: MFCD00239399
CAS Number: 19484-57-2
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/56/34/21285634.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR