3-Chloro-2-fluorophenylhydrazine hydrochloride
Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms:
(3-Chloro-2-fluorophenyl)hydrazine hydrochloride
2628685GM
100286-992EA
150.38
USD
100286-992
3-Chloro-2-fluorophenylhydrazine hydrochloride
3-Chloro-2-fluorophenylhydrazine hydrochloride
Formula:
C₆H₇Cl₂FN₂ MW: 197.0400 g/mol |
MDL Number:
MFCD03094280 CAS Number: 517920-75-1 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...