5-Chloro-2-fluoroaniline ≥98%
Supplier: INDOFINE CHEMICAL COMPANY MS
09322125GM
100273-232EA
104.62
USD
100273-232
5-Chloro-2-fluoroaniline ≥98%
5-Chloro-2-fluoroaniline
Formula:
C₆H₅ClFN MW: 145.56 g/mol Melting Pt: 20…22 °C |
MDL Number:
MFCD00069416 CAS Number: 2106-05-0 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...