1-Chloro-2,4-dibromo-6-fluorobenzene ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 2-Chloro-1,5-dibromo-3-fluorobenzene2-Chloro-3,5-dibromo-1-fluorobenzene1,5-Dibromo-2-chloro-3-fluorobenzene

263605GM
100294-018EA 202.69 USD
100294-018
1-Chloro-2,4-dibromo-6-fluorobenzene ≥98%
1-Chloro-2,4-dibromo-6-fluorobenzene
Formula: C₆H₂Br₂ClF
MW: 288.34 g/mol
MDL Number: MFCD00142891
CAS Number: 202925-04-0
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/72/86/21287286.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR