(R)-Methyl-2-amino-3-chloropropanoate hydrochloride ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: H-β-Chloro-Ala-OMe·HCl

0410361GM
100278-636EA 224.92 USD
100278-636
(R)-Methyl-2-amino-3-chloropropanoate hydrochloride ≥98%, powder
(R)-Methyl-2-amino-3-chloropropanoate hydrochloride

  • White powder
  • Store at 0-8 °C

Formula: C₄H₉Cl₂NO₂
MW: 174.021 g/mol
Melting Pt: 147…150 °C
MDL Number: MFCD00270526
CAS Number: 17136-54-8
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/65/03/21286503.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR