Chitin, powder
024068100GM
100263-638EA
129.46
USD
100263-638
Chitin, powder
Chitin
- Powder
- Store in a cool, dry place
Formula:
C₃₀H₅₀N₄O₁₉ |
MDL Number:
MFCD00466914 CAS Number: 1398-61-4 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...