Α-Chaconine ≥98% (by HPLC)

Supplier: INDOFINE CHEMICAL COMPANY MS

021553S5MG
100263-540EA 517.85 USD
100263-540
Α-Chaconine ≥98% (by HPLC)
Α-Chaconine
Formula: C₄₅H₇₃NO₁₄
Melting Pt: 240…253 °C
MDL Number: MFCD09039102
CAS Number: 20562-03-2
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/55/70/21285570.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR