Cedrol ≥98%, crystalline powder
Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms:
(3R,3aS,6R,7R,8aS)-3,6,8,8-Tetramethyloctahydro-1H-3a,7-methanoazulen-6-ol
025162S1GM
100269-986EA
372.69
USD
100269-986
Cedrol ≥98%, crystalline powder
Cedrol
- White crystalline powder
- Store at 2-8 °C
Formula:
C₁₅H₂₆O MW: 222.37 g/mol Melting Pt: 84.5 °C |
MDL Number:
MFCD00062952 CAS Number: 77-53-2 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...