Α-Cedrene with Gc ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

025432S100MG
100263-730EA 333.46 USD
100263-730
Α-Cedrene with Gc ≥98%
Α-Cedrene with Gc
Formula: C₁₅H₂₄
MDL Number: MFCD00063003
CAS Number: 469-61-4
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/55/80/21285580.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR