L-Hyp-(tBu)-OtBu·HCI ≥98%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: H-Hyp(tBu)-OtBu·HCl

0418261GM
100279-822EA 285.08 USD
100279-822
L-Hyp-(tBu)-OtBu·HCI ≥98%, powder
L-Hyp-(tBu)-OtBu·HCI

  • White powder
  • Store at 0-8 °C

Formula: C₁₃H₂₅NO₃·HCl
MW: 279.84 g/mol
Melting Pt: 140.4…140.7 °C
MDL Number: MFCD01862362
CAS Number: 367453-05-2
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/63/36/21286336.jpg https://us.vwr.com/stibo/web/std.lang.all/63/37/21286337.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR