H-Hyp(tBu)-OH ≥99.5%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

0418251GM
100279-820EA 122.92 USD
100279-820
H-Hyp(tBu)-OH ≥99.5%, powder
H-Hyp(tBu)-OH

  • White fine flowing powder
  • Store at 0-8 °C

Formula: C₉H₁₇NO₃
Melting Pt: 198…200 °C
MDL Number: MFCD00153445
CAS Number: 79775-07-8
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/63/35/21286335.jpg https://us.vwr.com/stibo/web/std.lang.all/63/34/21286334.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR