H-D-Cys(tBu)-OH·HCl ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (S)-2-Amino-3-(tert-butylthio)propanoic acid hydrochlorideH-D-Cys(tBu)-OH·HCl

049641GM
100295-302EA 303.38 USD
100295-302
H-D-Cys(tBu)-OH·HCl ≥98%
H-D-Cys(tBu)-OH·HCl
Formula: C₇H₁₆ClNO₂S
MW: 213.72 g/mol
MDL Number: MFCD00135154
CAS Number: 200353-65-7
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/74/53/21287453.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR