N-(2-Bromobenzyloxycarbonyloxy)succinimide ≥98%, solid

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 2-Bromobenzyl Succinimidyl CarbonateCarbonic Acid 2-Bromobenzyl Succinimidyl EsterZ(2-Br)-OSu

0494925GM
100293-026EA 105.92 USD
100293-026
N-(2-Bromobenzyloxycarbonyloxy)succinimide ≥98%, solid
N-(2-Bromobenzyloxycarbonyloxy)succinimide

  • White Solid
  • Store at 0-8 °C

Formula: C₁₂H₁₀BrNO₅
MW: 328.12 g/mol
Melting Pt: 107…109 °C
MDL Number: MFCD00082848
CAS Number: 128611-93-8
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/72/45/21287245.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR