4-Bromo-o-phenylenediamine ≥98%, crystalline solid

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 2-Amino-4-bromophenylamine4-Bromobenzene-1,2-diamine4-Bromo-1,2-phenylenediamine4-Bromo-1,2-diaminobenzene1-Bromo-3,4-diaminobenzene

250131GM
100267-872EA 39.23 USD
100267-872
4-Bromo-o-phenylenediamine ≥98%, crystalline solid
4-Bromo-o-phenylenediamine

  • Grey crystalline solid
  • Store in a cool, dry place

Formula: C₆H₇BrN₂
MW: 187.04 g/mol
Melting Pt: 58…60 °C
MDL Number: MFCD02660622
CAS Number: 1575-37-7
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/57/76/21285776.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR