3-Bromo-4-methoxybenzoic acid ≥99%, Powder
Supplier: INDOFINE CHEMICAL COMPANY MS
195851GM
100267-552EA
26.15
USD
100267-552
3-Bromo-4-methoxybenzoic acid ≥99%, Powder
3-Bromo-4-methoxybenzoic acid
- White powder
- Store in a cool, dry place
Formula:
C₈H₇BrO₃ MW: 231.05 g/mol Melting Pt: 220…222 °C |
MDL Number:
MFCD00020295 CAS Number: 99-58-1 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...