5'-Bromo-2'-hydroxypropiophenone ≥99%, Powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: 1-(5-Bromo-2-hydroxyphenyl)propan-1-one

195261GM
100267-454EA 111.15 USD
100267-454
5'-Bromo-2'-hydroxypropiophenone ≥99%, Powder
5'-Bromo-2'-hydroxypropiophenone

  • Light yellow shining powder
  • Store in a cool, dry place

Formula: C₉H₉BrO₂
MW: 229.0706 g/mol
Melting Pt: 75…77.2 °C
MDL Number: MFCD00218577
CAS Number: 17764-93-1
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/57/55/21285755.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR