3-Bromo-4-hydroxybenzoic acid ≥98%, Powder

Supplier: INDOFINE CHEMICAL COMPANY MS

191555GM
100269-216EA 36.62 USD
100269-216
3-Bromo-4-hydroxybenzoic acid ≥98%, Powder
3-Bromo-4-hydroxybenzoic acid

  • White powder
  • Store in a cool, dry place

Formula: C₇H₅BrO₃
MW: 217.02 g/mol
Melting Pt: 177…179 °C
MDL Number: MFCD00017547
CAS Number: 14348-41-5
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/58/92/21285892.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR