5-Bromo-2,4-difluoroaniline ≥98%

270375GM
100289-298EA 89.88 USD
100289-298
5-Bromo-2,4-difluoroaniline ≥98%
5-Bromo-2,4-difluoroaniline
Formula: C₆H₄BrF₂N
MW: 208.0000 g/mol
Melting Pt: 25…26 °C
MDL Number: MFCD03426368
CAS Number: 452-92-6
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/70/46/21287046.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR