Boc-Val-OMe

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: N-Boc-L-valine methyl esterBoc-Val-OMe(S)-Methyl 2-[(tert-Butoxycarbonyl)amino]-3-methylbutanoateN-Boc-L-valine methyll esterN-(tert-Butoxycarbonyl)-L-valine methyll ester

049385GM
100295-290EA 111.15 USD
100295-290
Boc-Val-OMe
Boc-Val-OMe
Formula: C₁₁H₂₁NO₄
MW: 231.292 g/mol
MDL Number: MFCD00672510
CAS Number: 58561-04-9
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/74/01/21287401.jpg https://us.vwr.com/stibo/web/std.lang.all/74/00/21287400.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR