Boc-S-trityl-D-cysteine ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: N-Boc-S-trityl-D-cysteineBoc-D-Cys(Trt)-OH

049021GM
100295-282EA 87.62 USD
100295-282
Boc-S-trityl-D-cysteine ≥99%
Boc-S-trityl-D-cysteine
Formula: C₂₇H₂₉NO₄S
MW: 463.592 g/mol
MDL Number: MFCD00236839
CAS Number: 87494-13-1
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR