Boc-D-His(Tos)-OH ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

048951GM
100281-970EA 160.85 USD
100281-970
Boc-D-His(Tos)-OH ≥99%
Boc-D-His(Tos)-OH
Formula: C₁₈H₂₃N₃O₆S
MDL Number: MFCD00037890
CAS Number: 69541-68-0
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/75/14/21287514.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR