Boc-Tic-OH ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (S)-N-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid(S)-2-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acidBoc-Tic-OHBoc-L-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid(S)-2-(tert-Butoxycarbonyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

048801GM
100295-278EA 30.08 USD
100295-278
Boc-Tic-OH ≥98%
Boc-Tic-OH
Formula: C₁₅H₁₉NO₄
MW: 277.32 g/mol
MDL Number: MFCD00143845
CAS Number: 78879-20-6
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/74/52/21287452.jpg https://us.vwr.com/stibo/web/std.lang.all/74/51/21287451.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR