N-Boc-D-prolinol ≥98% (by HPLC), Powder
Supplier: INDOFINE CHEMICAL COMPANY MS
Synonyms:
(R)-(+)-1-Boc-2-pyrrolidinemethanol
, (R)-1-Boc-2-pyrrolidinemethanol
, N-t-Boc-D-prolinol
, (R)-(+)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanol
, N-tert-Butoxycarbonyl-D-prolinol
, Boc-D-Prolinol
, (R)-1-(tert-Butoxycarbonyl)-2-pyrrolidinemethanol
048641GM
100281-924EA
34
USD
100281-924
N-Boc-D-prolinol ≥98% (by HPLC), Powder
N-Boc-D-prolinol
- White powder
- Store at 0-8 °C
Formula:
C₁₀H₁₉NO₃ MW: 201.3 g/mol Melting Pt: 52…65 °C |
MDL Number:
MFCD00040580 CAS Number: 83435-58-9 ADR: , |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...