Boc-D-Pro-OH ≥99% (by HPLC), Powder

Synonyms: N-Boc-D-proline(2R)-1-[(tert-butoxy)carbonyl]pyrrolidine-2-carboxylic acid

0485525GM
100277-428EA 28.89 USD
100277-428
Boc-D-Pro-OH ≥99% (by HPLC), Powder
Boc-D-Pro-OH

  • White to off-white powder
  • Store at 0-8 °C

Formula: C₁₀H₁₇NO₄
MW: 215.25 g/mol
Melting Pt: 132…140 °C
MDL Number: MFCD00063226
CAS Number: 37784-17-1
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/64/46/21286446.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR