Boc-PHG-OSu ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

048491GM
100281-894EA 151.69 USD
100281-894
Boc-PHG-OSu ≥99%
Boc-PHG-OSu
Formula: C₁₇H₂₀N₂O₆
MDL Number: MFCD00190826
CAS Number: 201152-47-8
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/67/65/21286765.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR