N-(tert-Butoxycarbonyl)-4-nitro-D-phenylalanine ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: N-Boc-4-nitro-D-phenylalanineBoc-D-Phe(4-NO2)-OH

048151GM
100281-842EA 115.08 USD
100281-842
N-(tert-Butoxycarbonyl)-4-nitro-D-phenylalanine ≥98%
N-(tert-Butoxycarbonyl)-4-nitro-D-phenylalanine
Formula: C₁₄H₁₈N₂O₆
MW: 310.31 g/mol
MDL Number: MFCD00069880
CAS Number: 61280-75-9
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/60/46/21286046.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR