Boc-D-Arg(NO₂)-OH ≥99%, powder

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: N2-(tert-Butoxycarbonyl)-Nw-nitro-D-arginine

048131GM
100274-282EA 141.23 USD
100274-282
Boc-D-Arg(NO₂)-OH ≥99%, powder
Boc-D-Arg(NO₂)-OH

  • White powder
  • Store at 0-8 °C

Formula: C₁₁H₂₁N₅O₆
MW: 319.318 g/mol
MDL Number: MFCD00038091
CAS Number: 50913-12-7
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/61/77/21286177.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR