N-(tert-Butoxycarbonyl)-3-(2-naphthyl)-L-alanine ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: (S)-alpha-(Boc-amino)-2-naphthalenepropionic Acid(S)-alpha-tert-Butoxycarbonylamino-2-naphthalenepropionic AcidBoc-2-Nal-OHN-Boc-3-(2-naphthyl)-L-alanine

048121GM
100277-404EA 138.62 USD
100277-404
N-(tert-Butoxycarbonyl)-3-(2-naphthyl)-L-alanine ≥99%
N-(tert-Butoxycarbonyl)-3-(2-naphthyl)-L-alanine
Formula: C₁₈H₂₁NO₄
MW: 315.37 g/mol
MDL Number: MFCD00079671
CAS Number: 58438-04-3
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/67/56/21286756.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR