Boc-3-(1-naphthyl)-L-alanine ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: N-Boc-3-(1-naphthyl)-L-alanineBoc-1-Nal-OH(S)-2-((tert-Butoxycarbonyl)amino)-3-(naphthalen-1-yl)propanoic acid

048111GM
100292-986EA 129.46 USD
100292-986
Boc-3-(1-naphthyl)-L-alanine ≥99%
Boc-3-(1-naphthyl)-L-alanine
Formula: C₁₈H₂₁NO₄
MW: 315.369 g/mol
MDL Number: MFCD00085377
CAS Number: 55447-00-2
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/74/50/21287450.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR