Boc-N-methyl-L-leucine ≥98%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: Boc-N-Me-Leu-OH

048021GM
100281-824EA 91.54 USD
100281-824
Boc-N-methyl-L-leucine ≥98%
Boc-N-methyl-L-leucine
Formula: C₁₂H₂₃NO₄
MW: 245.319 g/mol
MDL Number: MFCD00038522
CAS Number: 53363-89-6
ADR: ,
https://us.vwr.com/stibo/web/std.lang.all/75/12/21287512.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR