(Boc-Cys-OH)₂ ≥99%

Supplier: INDOFINE CHEMICAL COMPANY MS

Synonyms: N,N'-di-Boc-L-cystineAK-32714Bis-Boc-L-cystine

0462225GM
100277-296EA 163.46 USD
100277-296
(Boc-Cys-OH)₂ ≥99%
(Boc-Cys-OH)₂
Formula: C₁₆H₂₈N₂O₈S₂
MW: 440.526 g/mol
MDL Number: MFCD00038250
CAS Number: 10389-65-8
ADR: ,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR